Zinc acrylate
Supplier: Thermo Scientific Chemicals
041780-A1
041780-30
041780-18
AA41780-A1EA
454
USD
AA41780-A1
AA41780-30
AA41780-18
Zinc acrylate
Zinc acrylate
CAS No.: 14643-87-9
Molecular Formula: Zn(H2C=CHCO2)2
Formula Weight: 207.50
Physical Form: Powder
Density: 1.6
Melting Point: ≈250°
UN No.: 3077
MDL No.: MFCD00067208
Formula:
Zn(H₂C=CHCO₂)₂ |
MDL Number:
MFCD00067208 CAS Number: 14643-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...