Sulfur in Residual Oil Standard Solutions, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

041753-AE 041751-AE 041755-AE 041756-AE 041757-AE 041752-AE
AA41753-AEEA 245 USD
AA41753-AE AA41751-AE AA41755-AE AA41756-AE AA41757-AE AA41752-AE
Sulfur in Residual Oil Standard Solutions, Specpure®, Thermo Scientific
Standards Petroleum Industry Standards

Physical Form: Liquid
MDL No.: MFCD00197838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR