Potassium aquapentachlororuthenate(III) (≥26.4% Ru) ≥99.99% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
041716-06
041716-03
AA41716-06EA
263
USD
AA41716-06
AA41716-03
Potassium aquapentachlororuthenate(III) (≥26.4% Ru) ≥99.99% (metals basis), Premion®
Potassium aquapentachlororuthenate(III)
Purity: 99.99%
CAS No.: 14404-33-2
Molecular Formula: K2(RuCl5)(H2O)
Formula Weight: 374.55 (356.53anhy)
Physical Form: Crystalline
Appearance: Brown to red
MDL No.: MFCD00058747
Formula:
K₂(RuCl₅)(H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00058747 CAS Number: 14404-33-2 EINECS: 238-374-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...