Poly(styrene sulfonic acid) sodium salt, M.W. 70,000

Supplier: Thermo Scientific Chemicals

041688-18 041688-30
AA41688-18EA 82.95 USD
AA41688-18 AA41688-30
Poly(styrene sulfonic acid) sodium salt, M.W. 70,000
Poly(styrene sulfonic acid) sodium salt

CAS No.: 25704-18-1 Molecular Formula: [-CH2CH(C6H4)SO3Na)-]n Physical Form: Powder MDL No.: MFCD00084449

Formula: [-CH₂CH(C₆H₄)SO₃Na)-]n
Storage Temperature: Ambient
MDL Number: MFCD00084449
CAS Number: 25704-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR