Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ca(TMHD)2
041589-04
041589-09
AA41589-18EA
960.15
USD
AA41589-18
AA41589-04
AA41589-09
AA41589-02
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium ≥99.99% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium
Purity: 99.99%
CAS No.: 36818-89-0
Molecular Formula: C22H38CaO4
Formula Weight: 406.57
Physical Form: Powder
Sensitivity: Hygroscopic
Boiling Point: 205°/0.1mm subl.
MDL No.: MFCD00064757
Formula:
C₂₂H₃₈CaO₄ MW: 406.57 g/mol Boiling Pt: 205 °C (0.1 mmHg) (subl.) Melting Pt: 202…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064757 CAS Number: 36818-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...