Cobalt(II) Phthalocyanine

Supplier: Thermo Scientific Chemicals
Warning

041496-03 041496-06 041496-14
AA41496-03EA 37 USD
AA41496-03 AA41496-06 AA41496-14
Cobalt(II) Phthalocyanine
Cobalt(II) Phthalocyanine

Fieser: 9,119 10,102 11,138


CAS No.: 3317-67-7 Molecular Formula: C32H16N8Co Formula Weight: 571.47 Physical Form: Powder Appearance: Dark blue to purple Odor: Odorless MDL No.: MFCD00010718

Formula: C₃₂H₁₆CoN₈
Storage Temperature: Ambient
MDL Number: MFCD00010718
CAS Number: 3317-67-7
EINECS: 222-012-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR