Copper(II) ethylacetoacetate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

041489-09 041489-04 041489-18
AA41489-09EA 117 USD
AA41489-09 AA41489-04 AA41489-18
Copper(II) ethylacetoacetate ≥97%
Copper(II) ethylacetoacetate

Soluble in ethanol


Purity: 97% CAS No.: 14284-06-1 Molecular Formula: (CH3CH2CO2CHCOCH3)2Cu Formula Weight: 321.81 Physical Form: Powder Odor: Odorless Melting Point: 192° Solubility: Soluble in ethanol MDL No.: MFCD00017759

Formula: (CH₃CH₂CO₂CHCOCH₃)₂Cu
MW: 321.81 g/mol
Melting Pt: 192 °C
Storage Temperature: Ambient
MDL Number: MFCD00017759
CAS Number: 14284-06-1
EINECS: 238-181-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR