Tetramethylammonium hydrogen phthalate ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
041360-14
041360-22
AA41360-14EA
63
USD
AA41360-14
AA41360-22
Tetramethylammonium hydrogen phthalate ≥99%
Tetramethylammonium hydrogen phthalate
Applications: In electrolyte formulations
Purity: 99+%
CAS No.: 79723-02-7
Molecular Formula: C12H17NO4
Formula Weight: 239.27
Physical Form: Crystalline
Melting Point: 148-150°
UN No.: 2811
MDL No.: MFCD00216634
Applications: In electrolyte formulations
Formula:
C₁₂H₁₇NO₄ Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216634 CAS Number: 79723-02-7 EINECS: 416-900-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...