Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99%, medium molecular weight, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

041242-14 041242-22 041242-36
AA41242-14EA 40 USD
AA41242-14 AA41242-22 AA41242-36
Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99%, medium molecular weight, hydrolysed
Polyvinyl alcohol

Average M.W. 57,000-66,000


Purity: 98-99% CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Odor: Odorless MDL No.: MFCD00081922

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR