Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 20 mesh woven from 0.102mm (0.004in) Type 316

Supplier: Thermo Scientific Chemicals

040943-GT 040943-FL 040943-HB
AA40943-GTEA 107 USD
AA40943-GT AA40943-FL AA40943-HB
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 20 mesh woven from 0.102mm (0.004in) Type 316
Stainless steel

Open area: 84.6%; Width of opening: 0.046in


Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% Physical Form: Wire Cloth MDL No.: MFCD00148499

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR