Germanium-n-butoxide ≥97%

Supplier: Thermo Scientific Chemicals
Warning

040816-06 040816-03 040816-14
AA40816-06EA 140 USD
AA40816-06 AA40816-03 AA40816-14
Germanium-n-butoxide ≥97%
Germanium-n-butoxide

Applications: Oxide films


Applications: Oxide films


Applications: Oxide films

Formula: C₁₆H₃₆GeO₄
MW: 365.05 g/mol
Boiling Pt: 143 °C (8 mmHg)
Density: 1,017
Flash Pt: 112 °C (234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015347
CAS Number: 25063-27-8
EINECS: 246-595-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR