Strontium-2-ethylhexanoate 40% (8 - 12% Sr) in 2-ethylhexanoic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethylhexanoic acid strontium salt

040719-22 040719-14
AA40719-22EA 636 USD
AA40719-22 AA40719-06 AA40719-14
Strontium-2-ethylhexanoate 40% (8 - 12% Sr) in 2-ethylhexanoic acid
Strontium 2-ethylhexanoate

CAS No.: 2457-02-5 Molecular Formula: C16H30O4Sr Formula Weight: 374.04 Physical Form: Liquid Odor: Mild MDL No.: MFCD00070438

Formula: C₁₆H₃₀O₄Sr
Storage Temperature: Ambient
MDL Number: MFCD00070438
CAS Number: 2457-02-5
EINECS: 219-536-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR