Samarium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

040567-09 040567-18
AA40567-09EA 94.6 USD
AA40567-09 AA40567-18
Samarium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Samarium (III) perchlorate

CAS No.: 13569-60-3 Molecular Formula: Sm(ClO4)3.6H2O Formula Weight: 556.80 (448.70anhy) Physical Form: Liquid Odor: Odorless UN No.: 3211 MDL No.: MFCD00049769

Formula: Cl₃O₁₂Sm
MW: 448.698 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049769
CAS Number: 13569-60-3
EINECS: 236-986-6
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR