Gadolinium(III) hydroxide hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

040507-06 040507-14
AA40507-06EA 59.6 USD
AA40507-06 AA40507-14
Gadolinium(III) hydroxide hydrate ≥99.9% (REO, rare earth oxide basis)
Gadolinium(III) hydroxide hydrate

Purity: 99.9% CAS No.: 100634-91-1 Molecular Formula: Gd(OH)3.xH2O Formula Weight: 208.27(anhy) Physical Form: Powder MDL No.: MFCD00049930

Formula: Gd(OH)₃.xH₂O
MW: 208.27 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00049930
CAS Number: 100634-91-1
EINECS: 240-516-5
Merck Index: 14,4326

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR