Cerium(IV)-2-methoxyethoxide 18 - 20% (w/w) in 2-methoxyethanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
040471-14
040471-22
040471-06
AA40471-14EA
339
USD
AA40471-14
AA40471-22
AA40471-06
Cerium(IV)-2-methoxyethoxide 18 - 20% (w/w) in 2-methoxyethanol
Cerium(IV)-2-methoxyethoxide
CAS No.: 876107-33-4
Molecular Formula: C12H28CeO8
Formula Weight: 440.47
Physical Form: Liquid
Density: 1.02
Sensitivity: Moisture Sensitive
Appearance: Yellow
UN No.: 1188
MDL No.: MFCD00236530
Formula:
C₁₂H₂₈CeO₈ Density: 1.02 Storage Temperature: Ambient |
MDL Number:
MFCD00236530 CAS Number: 876107-33-4 UN: 1188 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...