Hydrogen tetrabromoaurate(III) hydrate (≥32% Au) ≥99.99% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bromoauric acid
040433-06
040433-03
AA40433-06EA
550
USD
AA40433-06
AA40433-03
Hydrogen tetrabromoaurate(III) hydrate (≥32% Au) ≥99.99% (metals basis), Premion®
Hydrogen tetrabromoaurate(III) hydrate
Soluble in water and alcohol
Purity: 99.99%
Molecular Formula: HAuBr4.xH2O (x≈5)
Formula Weight: 517.61(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: ca 27°
Solubility: Soluble in water and alcohol
UN No.: 3260
MDL No.: MFCD00054118
Formula:
HAuBr₄·xH₂O MW: 517.61 g/mol (anhy) Melting Pt: ∼ 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150570 CAS Number: 17083-68-0 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...