Cerium molybdenum oxide ≥99%
Supplier: Thermo Scientific Chemicals
040210-22
040210-06
040210-14
AA40210-22EA
1171
USD
AA40210-22
AA40210-06
AA40210-14
Cerium molybdenum oxide ≥99%
Cerium molybdenum oxide
Purity: 99%
CAS No.: 13454-70-1
Molecular Formula: Ce2(MoO4)3
Formula Weight: 760.05
Physical Form: Powder
Odor: Odorless
Melting Point: 973°
MDL No.: MFCD00798520
Formula:
Ce₂(MoO₄)₃ MW: 760.05 g/mol Melting Pt: 973 °C Storage Temperature: Ambient |
MDL Number:
MFCD00798520 CAS Number: 13454-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...