Silver tungsten oxide ≥99%
Supplier: Thermo Scientific Chemicals
Insoluble in water. Soluble in potassium cyanide, ammonium hydroxide and nitric acid
Purity: 99%
CAS No.: 13465-93-5
Molecular Formula: Ag2O4W
Formula Weight: 463.58
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Light yellow
Odor: Odorless
Solubility: Insoluble in water. Soluble in potassium cyanide, ammonium hydroxide and nitric acid
MDL No.: MFCD00014147
Formula:
Ag₂O₄W Storage Temperature: Ambient |
MDL Number:
MFCD00014147 CAS Number: 13465-93-5 EINECS: 236-708-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...