Samarium(III) hydroxide hydrate ≥99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

039512-06 039512-14
AA39512-06EA 53.7 USD
AA39512-06 AA39512-14
Samarium(III) hydroxide hydrate ≥99% (REO, rare earth oxide basis)
Samarium(III) hydroxide hydrate

Purity: 99% CAS No.: 20403-06-9 Molecular Formula: Sm(OH)3.xH2O Formula Weight: 201.37(anhy) Physical Form: Lump/Powder UN No.: 3262 MDL No.: MFCD00050108

Formula: Sm(OH)₃·xH₂O
MW: 201.37 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00050108
CAS Number: 20403-06-9
EINECS: 243-787-8
UN: 3262
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR