Magnesium niobium oxide ≥99.9%

Supplier: Thermo Scientific Chemicals

039373-09 039373-18 039373-30
AA39373-09EA 102 USD
AA39373-09 AA39373-18 AA39373-30
Magnesium niobium oxide ≥99.9%
Magnesium niobium oxide

Purity: 99.9% CAS No.: 12163-26-7 Molecular Formula: MgNb2O6 Formula Weight: 306.11 Physical Form: -200 Mesh Powder Odor: Odorless MDL No.: MFCD00210631

Formula: MgNb₂O₆
MW: 306.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210631
CAS Number: 12163-26-7
EINECS: 235-310-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR