Lithium tartrate monohydrate ≥99%

Supplier: Thermo Scientific Chemicals

039357-30 039357-18
AA39357-30EA 228 USD
AA39357-30 AA39357-18
Lithium tartrate monohydrate ≥99%
Lithium tartrate monohydrate

Soluble in water


Purity: 99% CAS No.: 6108-32-3 Molecular Formula: C4H4Li2O6.H2O Formula Weight: 179.97 (161.97anhy) Physical Form: Powder/Aggregates Odor: Odorless Solubility: Soluble in water MDL No.: MFCD00054421

Formula: C₄H₄Li₂O₆·H₂O
MW: 179.97 g/mol (161.97 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00054421
CAS Number: 6108-32-3
EINECS: 212-772-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR