Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
039303-03
039303-MD
AA39303-03EA
142
USD
AA39303-03
AA39303-MD
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer
Purity: 99%
CAS No.: 12132-04-6
Molecular Formula: C24H30Mo2O4
Formula Weight: 574.38
Physical Form: Crystalline
Melting Point: 219° dec.
UN No.: 3466
MDL No.: MFCD00058846
Formula:
C₂₄H₃₀Mo₂O₄ Melting Pt: 219 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00058846 CAS Number: 12132-04-6 UN: 3466 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...