Iodotrimethylplatinum(IV)
Supplier: Thermo Scientific Chemicals
|
Warning
|
039286-02
039286-03
AA39286-02EA
237
USD
AA39286-02
AA39286-03
Iodotrimethylplatinum(IV)
Iodotrimethylplatinum(IV)
CAS No.: 14364-93-3
Molecular Formula: C3H9IPt
Formula Weight: 367.09
Physical Form: Crystalline
Sensitivity: Air and Light Sensitive
Appearance: White to pale orange
Odor: Odorless
MDL No.: MFCD00013702
Formula:
C₃H₉IPt Storage Temperature: Ambient |
MDL Number:
MFCD00013702 CAS Number: 14364-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...