Di-µ-chlorodichlorobis(ethylene)diplatinum(II) ≥97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zeise's dimer, Di-mu-chloro-dichlorobis(ethylene)diplatinum(II), Di-μ-chlorodichlorobis(ethylene)diplatinum(II)

039285-MD 039285-03
AA39285-MDEA 124 USD
AA39285-MD AAA39285-03
Di-µ-chlorodichlorobis(ethylene)diplatinum(II) ≥97%
Di-µ-chlorodichlorobis(ethylene)diplatinum(II)

Purity: 97% CAS No.: 12073-36-8 Molecular Formula: C4H8Cl4Pt Formula Weight: 588.10 Physical Form: Crystalline MDL No.: MFCD00058713

Formula: C₄H₄Cl₄Pt₂
MW: 584.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058713
CAS Number: 12073-36-8
EINECS: 235-135-6

Order Now

Specification Test Results

Form Crystalline

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR