Benzyldiphenylphosphine ≥99%
Supplier: Thermo Scientific Chemicals
039267-02
039267-09
039267-04
AA39267-02EA
29.3
USD
AA39267-02
AA39267-09
AA39267-04
Benzyldiphenylphosphine ≥99%
Benzyldiphenylphosphine
Purity: 99%
CAS No.: 7650-91-1
Molecular Formula: C6H5CH2P(C6H5)2
Formula Weight: 276.32
Physical Form: Powder
Sensitivity: Air Sensitive
Melting Point: 74-75°
MDL No.: MFCD00014083
Formula:
C₁₉H₁₇P MW: 276.319 g/mol Melting Pt: 74…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014083 CAS Number: 7650-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...