Iron(III) hexacyanoferrate(II)

Supplier: Thermo Scientific Chemicals

039259-14 039259-22
AA39259-14EA 70 USD
AA39259-14 AA39259-22
Iron(III) hexacyanoferrate(II)
Iron(III) hexacyanoferrate(II)

CAS No.: 14038-43-8 Molecular Formula: C18Fe7N18 Formula Weight: 859.25 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Dark blue Odor: Odorless MDL No.: MFCD00135663

Formula: C₁₈Fe₇N₁₈
Storage Temperature: Ambient
MDL Number: MFCD00135663
CAS Number: 14038-43-8
EINECS: 237-875-5
Merck Index: 14,7910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR