Magnesium(II) phthalocyanine ≥90%, tech.

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium phthalocyanine, Phthalocyanine Magnesium(II)

038601-03 038601-06
AA38601-03EA 52.9 USD
AA38601-03 AA38601-06
Magnesium(II) phthalocyanine ≥90%, tech.
Magnesium(II) phthalocyanine

Purity: 90% CAS No.: 1661-03-6 Molecular Formula: C32H16MgN8 Formula Weight: 536.84 Physical Form: Powder MDL No.: MFCD00041953

Formula: C₃₂H₁₆MgN₈
MW: 536.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041953
CAS Number: 1661-03-6
EINECS: 216-766-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR