Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

038367-22 038367-36
AA38367-22EA 54.12 USD
AA38367-22 AA38367-36
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
Cerium(III) ammonium nitrate tetrahydrate

CAS No.: 13083-04-0 Molecular Formula: H8CeN7O15.4H2O Formula Weight: 558.28 (486.22anhy) Physical Form: Crystalline Appearance: White UN No.: 1477 MDL No.: MFCD00210608

Formula: H₈CeN₇O₁₅·4H₂O
MW: 558.28 g/mol (486.22 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00210608
CAS Number: 13083-04-0
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR