Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced

Supplier: Thermo Scientific Chemicals

038292-09 038292-18
AA38292-09EA 92.9 USD
AA38292-09 AA38292-18
Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced
Palladium

Sold on a dry weight basis. Unit weight excludes water weight


Physical Form: 900-1100 m²/g

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR