Titanium(III) chloride aluminum(III) chloride (76.0 - 78.5% TiCl₃)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Titanium(III) chloride, Al reduced, Titanium aluminum chloride, Titanium(III) chloride, Aluminum titanium chloride

036729-36 036729-22
AA36729-36EA 584.46 USD
AA36729-36 AA36729-22
Titanium(III) chloride aluminum(III) chloride (76.0 - 78.5% TiCl₃)
Titanium(III) chloride aluminum(III) chloride

Activity: 165 min Catalyst for production of polypropylene


CAS No.: 12003-13-3 Molecular Formula: (TiCl3)3AlCl3 Formula Weight: 596.15 Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: 3260 MDL No.: MFCD00145412

Formula: (TiCl₃)₃AlCl₃
MW: 596.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145412
CAS Number: 12003-13-3
EINECS: 234-421-8
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR