CLP Protocol Test Kit for ICP Analysis, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

035259-AP 035419-BG 035421-AE 035422-AE 041331-AE 035420-AE 035256-AP 035417-BG 035418-BG 041330-AE
AA35259-APEA 72.8 USD
AA35259-AP AA35406-KS AA35419-BG AA35421-AE AA35422-AE AA41331-AE AA35420-AE AA35420-AP AA35256-AP AA35417-BG AA35418-BG AA41330-AE
CLP Protocol Test Kit for ICP Analysis, Specpure®, Thermo Scientific
Standards Cationic Standards Multi-Element Standards

This LabPak™ comes with all the necessary standard concentrates needed for the day to day operations to complete the EPA established CLP Protocol requirements. Minimal mixing and dilution of the standard concentrates make it easy by saving you time and money in preparing all the necessary CLP Protocol Standards. This LabPak™ contains the following CLP Protocol Test Kits for analysis by ICP:


UN No.: 3264
MDL No.: MFCD00146473


This LabPak™ contains the following CLP Protocol Test Kits for analysis by ICP:


Información para pedidos: Instrument Calibration (ICAL) Standards Kit (35417)
Initial Calibration Verification (ICV) Standards Kit (35418)
Continuing Calibration Verification (CCV) Standards Kit (35419)
Contract Required Detection Limits (CRDL) Standard (35420)
Interference Check Standard A (ICS-A) Solution (35256) - (2 each)
Interference Check Standard AB-1 (ICS-AB1) Solution (35421)
Interference Check Standard AB-2 (ICS-AB2) Solution (41330)
Water Spiking Standard A (WSS-A) Solution (35422)
Water Spiking Standard B (WSS-B) Solution (41331)
Initial Calibration Blank/Continuing Calibration Blank 4 (ICB/CCB) Standards Kit (35259) - (4 each)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR