o-Carborane ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
035131-04
035131-09
AA35131-04EA
230
USD
AA35131-04
AA35131-09
o-Carborane ≥98%
o-Carborane
Insoluble in water
Purity: 98%
CAS No.: 16872-09-6
Molecular Formula: C2H12B10
Formula Weight: 144.25
Physical Form: Powder
Density: 0.95
Appearance: White
Melting Point: 285-287°
Solubility: Insoluble in water
UN No.: 1325
MDL No.: MFCD00012169
Formula:
C₂H₂B₁₀ MW: 144.23 g/mol Melting Pt: 285…287 °C Density: 0.95 Storage Temperature: Ambient |
MDL Number:
MFCD00012169 CAS Number: 16872-09-6 EINECS: 240-897-8 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...