Sodium borodeuteride (99% D) (Isotopic), rods

Supplier: Thermo Scientific Chemicals
Danger

035102-03 035102-06
AA35102-03EA 90.42 USD
AA35102-03 AA35102-06
Sodium borodeuteride (99% D) (Isotopic), rods
Sodium borodeuteride

D 18%


Purity: 99% CAS No.: 15681-89-7 Molecular Formula: BD4Na Formula Weight: 41.88 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White pH: 11 Melting Point: ≈400° dec. UN No.: 1426 MDL No.: MFCD00003519

Formula: D₄BNa
Melting Pt: ∼400 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003519
CAS Number: 15681-89-7
EINECS: 239-764-7
UN: 1426
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR