(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2S,4S)-Pentane-2,4-diylbis(diphenylphosphine), (S,S)-BDPP

033668-MD 033668-03
AA33668-MDEA 90 USD
AA33668-MD AA33668-03
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane ≥99%
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane

Purity: 99% CAS No.: 77876-39-2 Molecular Formula: C29H30P2 Formula Weight: 440.50 Physical Form: Crystalline MDL No.: MFCD00058923

Formula: C₂₉H₃₀P₂
MW: 440.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058923
CAS Number: 77876-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR