2-Phenylisobutyric acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-2-phenylpropanoic acid, α,α-Dimethylphenylacetic acid

032884-04 032884-09
AA32884-04EA 119 USD
AA32884-04 AA32884-09
2-Phenylisobutyric acid
2-Phenylisobutyric acid

Insoluble in water


CAS No.: 826-55-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Physical Form: Crystalline Melting Point: 80-82° Boiling Point: 146.5° Solubility: Insoluble in water MDL No.: MFCD00014332

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Boiling Pt: 146.5 °C
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00014332
CAS Number: 826-55-1
EINECS: 212-556-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR