Calcium-2,4-pentanedionate hydrate ≥99%
Supplier: Thermo Scientific Chemicals
030526-09
030526-18
AA30526-30EA
208.68
USD
AA30526-30
AA30526-09
AA30526-18
Calcium-2,4-pentanedionate hydrate ≥99%
Calcium-2,4-pentanedionate hydrate
Purity: 99%
CAS No.: 345909-31-1
Molecular Formula: C10H14CaO4.xH2O
Formula Weight: 238.30(anhy)
Physical Form: Crystalline
Melting Point: 175° dec.
MDL No.: MFCD00013486
Formula:
C₁₀H₁₄CaO₄.xH₂O MW: 238.30 g/mol (anhy) Melting Pt: 175 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013486 CAS Number: 345909-31-1 EINECS: 243-001-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...