Triethyl-2-phosphonoppentanoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-(diethoxyphosphoryl)pentanoate
030413-04
030413-09
AA30413-04EA
125
USD
AA30413-04
AA30413-02
AAA30413-09
Triethyl-2-phosphonoppentanoate 98%
Triethyl-2-phosphonoppentanoate
Purity: 98%
CAS No.: 35051-49-1
Molecular Formula: C11H23O5P
Formula Weight: 266.27
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00015163
Formula:
C₁₁H₂₃O₅P MW: 266.27 g/mol Boiling Pt: 98 °C (2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00015163 CAS Number: 35051-49-1 |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...