Diiron boride 98%
Supplier: Thermo Scientific Chemicals
022952-14
022952-22
AA22952-14EA
141
USD
AA22952-14
AA22952-22
Diiron boride 98%
Diiron boride
Purity: 98%
CAS No.: 12006-85-8
Molecular Formula: Fe2B
Formula Weight: 122.50
Physical Form: -35 Mesh
Odor: Odorless
MDL No.: MFCD00016092
Formula:
BFe₂ Storage Temperature: Ambient |
MDL Number:
MFCD00016092 CAS Number: 12006-85-8 EINECS: 234-490-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...