Manganese(II) methoxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
Soluble in methanol
CAS No.: 7245-20-7
Molecular Formula: C2H6MnO2
Formula Weight: 117.01
Physical Form: Dark powder
Sensitivity: Moisture Sensitive
Solubility: Soluble in methanol
UN No.: 3181
MDL No.: MFCD00061476
Formula:
C₂H₆MnO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00061476 CAS Number: 7245-20-7 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...