Sodium metatungstate monohydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate

022534-30 022534-56 022534-18
AA22534-30EA 364.41 USD
AA22534-30 AAA22534-56 AA22534-18
Sodium metatungstate monohydrate
Sodium metatungstate monohydrate

CAS No.: 12141-67-2 Molecular Formula: Na6O39W12.H2O Formula Weight: 2986.13 (2968.12anhy) Physical Form: Powder Density: 5.47 Appearance: White Odor: Odorless MDL No.: MFCD00061466

Formula: Na₆O₃₉W₁₂·H₂O
MW: 2986.13 g/mol (2968.12 anhy)
Density: 5.47
Storage Temperature: Ambient
MDL Number: MFCD00061466
CAS Number: 12141-67-2
EINECS: 412-770-9

Order Now

Specification Test Results

Form Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR