Ethylenediaminetetraacetic acid disodium zinc salt, hydrate

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium zinc ethylenediaminetetraacetate, Ethylenediaminetetraacetic Acid Disodium Zinc Salt, Ethylenediaminetetraacetic acid zinc disodium salt

022426-22 022426-14 022426-36
AA22426-22EA 98.8 USD
AA22426-22 AA22426-14 AA22426-36
Ethylenediaminetetraacetic acid disodium zinc salt, hydrate
Ethylenediaminetetraacetic acid disodium zinc salt

CAS No.: 39208-16-7 Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2)2Zn.xH2O Formula Weight: 399.50(anhy) Physical Form: Powder MDL No.: MFCD00135187

Formula: (NaO₂CCH₂)₂NCH₂CH₂N(CH₂CO₂)₂Zn.xH₂O
MW: 399.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135187
CAS Number: 39208-16-7
EINECS: 237-865-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR