Ammonium tetrathiotungstate
Supplier: Thermo Scientific Chemicals
|
Warning
|
022373-09
022373-04
AA22373-09EA
362
USD
AA22373-09
AA22373-04
Ammonium tetrathiotungstate
Ammonium tetrathiotungstate
CAS No.: 13862-78-7
Molecular Formula: (NH4)2WS4
Formula Weight: 348.07
Physical Form: Chunks/Powder
Sensitivity: Hygroscopic
Odor: Like rotten eggs
MDL No.: MFCD00061410
Formula:
H₈N₂S₄W Storage Temperature: Ambient |
MDL Number:
MFCD00061410 CAS Number: 13862-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...