Diphenyldithiophosphonic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenylphosphinodithioic acid

017758-04 017758-09 017758-02
AA17758-04EA 149 USD
AA17758-04 AA17758-09 AA17758-02
Diphenyldithiophosphonic acid
Diphenyldithiophosphonic acid

CAS No.: 1015-38-9 Molecular Formula: (C6H5)2P(S)SH Formula Weight: 250.31 Physical Form: Crystalline Melting Point: 55-56° MDL No.: MFCD00014457

Formula: (C₆H₅)₂P(S)SH
MW: 250.31 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00014457
CAS Number: 1015-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR