Dysprosium(III) acetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

014595-14 014595-22
AA14595-14EA 61.6 USD
AA14595-14 AA14595-22
Dysprosium(III) acetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium (III) acetate tetrahydrate

Purity: 99.9% CAS No.: 15280-55-4 Molecular Formula: Dy(OOCCH)3.4H2O Formula Weight: 411.68 (339.62anhy) Physical Form: Crystalline Aggregates Odor: Odorless MDL No.: MFCD00013033

Formula: C₆H₉DyO₆·4H₂O
MW: 411.68 g/mol
Melting Pt: 120 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013033
CAS Number: 15280-55-4
EINECS: 242-565-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR