Scandium(III) carbonate hydrate ≥99.99%, REacton®
Supplier: Thermo Scientific Chemicals
013646-03
013646-06
AA13646-03EA
97
USD
AA13646-03
AA13646-06
Scandium(III) carbonate hydrate ≥99.99%, REacton®
Scandium(III) carbonate hydrate
Purity: 99.99%
Molecular Formula: Sc2(CO3)3.xH2O
Formula Weight: 269.94(anhy)
Physical Form: Crystalline
Melting Point:
Formula:
Sc₂(CO₃)₃·xH₂O MW: 269.94 g/mol (anhy) Melting Pt: >500 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149955 CAS Number: 5809-49-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...