Nonacarbonyldiiron ≥99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diiron nonacarbonyl, Iron nonacarbonyl

013062-09 013062-18
AA13062-09EA 106 USD
AA13062-09 AA13062-18
Nonacarbonyldiiron ≥99%
Nonacarbonyldiiron

Purity: 99% CAS No.: 15321-51-4 Molecular Formula: C9Fe2O9 Formula Weight: 363.79 Physical Form: Crystalline Density: 2.85 Sensitivity: Air Sensitive Appearance: Orange Melting Point: ca 100° dec. UN No.: 3179 MDL No.: MFCD00151465

Formula: C₉Fe₂O₉
MW: 363.78 g/mol
Melting Pt: ∼ 100 °C (dec.)
Density: 2,85
Storage Temperature: Ambient
MDL Number: MFCD00151465
CAS Number: 15321-51-4
EINECS: 239-359-5
UN: 3179
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR