Yttrium aluminum ≤1% ≥99.9% (metals basis excluding Ca), lumps
Supplier: Thermo Scientific Chemicals
012949-14
012949-22
AA12949-14EA
113
USD
AA12949-14
AA12949-22
Yttrium aluminum ≤1% ≥99.9% (metals basis excluding Ca), lumps
Yttrium aluminum
Purity: 99.9%
Molecular Formula: Y:Al; 87:12 wt%
MDL No.: MFCD01074900
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...