Tungsten boride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
012559-22
012559-14
AA12559-22EA
520
USD
AA12559-22
AA12559-14
Tungsten boride ≥99% (metals basis)
Tungsten boride
Purity: 99%
CAS No.: 12007-09-9
Molecular Formula: WB
Formula Weight: 194.66
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00049708
Formula:
BW Storage Temperature: Ambient |
MDL Number:
MFCD00049708 CAS Number: 12007-09-9 EINECS: 234-498-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...