Strontium acetate hemihydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
Synonyms:
Strontium Acetate Hemihydrate
012203-A1
012203-30
AA12203-A1EA
183
USD
AA12203-A1
AA12203-30
Strontium acetate hemihydrate, Reagent Grade
Strontium acetate hemihydrate
Converts to SrCO₃ on ignition
Soluble in water
CAS No.: 14692-29-6
Molecular Formula: Sr(OOCCH3)2.0.5H2O
Formula Weight: 214.72 (205.71anhy)
Physical Form: Crystalline
Melting Point: 150° -0.5H2O
Solubility: Soluble in water
MDL No.: MFCD00198064
Formula:
Sr(OOCCH₃)₂·0.5H₂O MW: 429.43 g/mol Melting Pt: 150 °C (-0.5H₂O) Density: 2.099 Storage Temperature: Ambient |
MDL Number:
MFCD00198064 CAS Number: 14692-29-6 EINECS: 208-854-8 Merck Index: 14,8836 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...