Chromium nitride

Supplier: Thermo Scientific Chemicals
Warning

012149-22 012149-14
AA12149-22EA 205 USD
AA12149-22 AA12149-14
Chromium nitride
Chromium nitride

CAS No.: 12053-27-9 Molecular Formula: Mixture of Cr2N and CrN Physical Form: -325 Mesh Powder Odor: Odorless MDL No.: MFCD00016048

Formula: Mixture of Cr₂N and CrN
Storage Temperature: Ambient
MDL Number: MFCD00016048
CAS Number: 12053-27-9
EINECS: 235-002-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR